Learn More
SiliCycle™ E-PAK™ Cartridges, Commercial Scale, SiliaFlash™ 60 - 200 μm, 60 Å
E-PAK™ Cartridges, Commercial Scale, SiliaFlash™ 60 - 200 μm, 60 Å irregular silica gels, F60, are conveniently packaged in a cartridge.
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
01-182-024
|
SiliCycle™
CSR10040B1650 |
50 x 16.5 cm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
01-182-023
|
SiliCycle™
CSR10040B16100 |
100 x 16.5 cm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Narrow particle size distribution
- Superior resolution
- Suitable for use with complex matrices
- Suitable for organic molecules (MW <1000 Dalton)
- Testing has shown a good correlation between the performance of lab, pilot, and commercial size cartridges.
Specifications
E-PAK Cartridge | |
Irregular |