Learn More
SiliCycle™ E-PAK™ Cartridges, Lab Scale, SiliaCarb™ C-CA Activated Carbon
E-PAK™ Cartridges, Lab Scale, SiliaCarb™ C-CA Activated Carbon is a chemically activated wood based powdered activated carbon conveniently packaged in a cartridge. E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
Supplier:  SiliCycle™ LSCCA51010
 View more versions of this product
Catalog No. 01-182-033
Add to Cart
Description
Description
  • Testing with loose media can be done with samples as small as a few milliliters and are normally done in advance of cartridge testing to identify the formula with the highest capacity to remove contaminants with the least loss of product.
  • E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
  • Testing has shown good correlation between the performance of lab, pilot, and commercial size cartridges.
  • Lab cartridges can be operated with pump or small pressure vessel.
Specifications
Specifications
Lab Scale E-PAK Cartridge
Black
10 Pk.
Chemically Activated Carbon
Removal of precious metal catalysts, pigments and reaction contaminants
10 x 5 cm
6 to 8
50 g