Learn More
SiliCycle™ E-PAK™ Cartridges, Lab Scale, SiliaCarb™ C-VW Activated Carbon
E-PAK™ Cartridges, Lab Scale, SiliaCarb™ C-VW Activated Carbon is a virgin wood based catalytic activated carbon conveniently packaged in a cartridge. E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
Supplier: SiliCycle™ LSCVW5255
Description
- Testing with loose media can be done with samples as small as a few milliliters and are normally done in advance of cartridge testing to identify the formula with the highest capacity to remove contaminants with the least loss of product.
- E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
- Testing has shown good correlation between the performance of lab, pilot, and commercial size cartridges.
- Lab cartridges can be operated with pump or small pressure vessel.
Specifications
Lab Scale E-PAK Cartridge | |
Black | |
5 Pk. | |
Virgin Wood Carbon |
Removal of precious metal catalysts, pigments and reaction contaminants | |
25 x 5 cm | |
6 to 8 | |
125 g |