Learn More
SiliCycle™ E-PAK™ Cartridges, Lab Scale, SiliaFlash™ Irregular Silica Gel F60, 40 to 63 μm, 60 Å
E-PAK™ Cartridges, Lab Scale, SiliaFlash™ Irregular Silica Gel F60, 40 - 63 μm, 60 Å is conveniently packaged in a cartridge. E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
Supplier: SiliCycle™ LSR10030B515
Description
- Narrow particle size distribution
- Superior resolution
- Suitable for use with complex matrices
- Suitable for organic molecules (MW <1000 Dalton)
- E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
- Testing has shown a good correlation between the performance of lab, pilot, and commercial size cartridges.
- Lab cartridges can be operated with a peristaltic pump or a small pressure vessel.
Specifications
Lab Scale E-PAK Cartridge | |
1 x 5 cm | |
40 to 63 μm | |
Bare Silica (Silicon Dioxyde, SiO2) |
Removal of various polar compounds | |
5 Pk. | |
60 Å | |
8 g |