Learn More
SiliCycle™ E-PAK™ Cartridges, Lab Scale, SiliaFlash™ Irregular Silica Gel G60, 60 - 200 μm, 60 Å
E-PAK™ Cartridges, Lab Scale, SiliaFlash™ Irregular Silica Gel G60, 60 - 200 μm, 60 Å is conveniently packaged in a cartridge. E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
Supplier:  SiliCycle™ LSR10040B5103
 View more versions of this product
Catalog No. 01-182-081
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Suitable for very dirty purification
  • Easier to handle
  • Suitable for organic molecules (MW <1000 Dalton)
  • E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
  • Testing has shown a good correlation between the performance of lab, pilot, and commercial size cartridges.
  • Lab cartridges can be operated with a peristaltic pump or a small pressure vessel.
Specifications
Specifications
Purification Cartridge
3 Pk.
60 Å
10 x 5 cm
60 to 200 μm
75 g