Learn More
SiliCycle™ E-PAK™ Cartridges, Lab Scale, SiliaFlash™ Irregular Silica Gel G60, 60 - 200 μm, 60 Å
E-PAK™ Cartridges, Lab Scale, SiliaFlash™ Irregular Silica Gel G60, 60 - 200 μm, 60 Å is conveniently packaged in a cartridge. E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
Supplier: SiliCycle™ LSR10040B5253
Description
- Suitable for very dirty purification
- Easier to handle
- Suitable for organic molecules (MW <1000 Dalton)
- E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
- Testing has shown a good correlation between the performance of lab, pilot, and commercial size cartridges.
- Lab cartridges can be operated with a peristaltic pump or a small pressure vessel.
Specifications
Purification Cartridge | |
3 Pk. | |
60 Å |
25 x 5 cm | |
60 to 200 μm | |
75 g |