Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SiliCycle™ E-PAK™ Cartridges, Lab Scale, SiliaFlash™ Irregular Silica Gel G60, 60 - 200 μm, 60 Å
E-PAK™ Cartridges, Lab Scale, SiliaFlash™ Irregular Silica Gel G60, 60 - 200 μm, 60 Å is conveniently packaged in a cartridge. E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
Specifications
Type | Purification Cartridge |
---|---|
Particle Size | 60 to 200 μm |
Pore Size | 60 Å |
Sorbent Weight | 75 g |
Catalog Number | Mfr. No. | Dimensions (L x W) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Quantity | Price | Quantity & Availability | ||||
01-182-079
|
SiliCycle™
LSR10040B510 |
10 x 5 cm | 1 Pk. | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
01-182-081
|
SiliCycle™
LSR10040B5103 |
10 x 5 cm | 3 Pk. | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
01-182-082
|
SiliCycle™
LSR10040B5105 |
10 x 5 cm | 5 Pk. | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
01-182-080
|
SiliCycle™
LSR10040B51010 |
10 x 5 cm | 10 Pk. | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
01-182-083
|
SiliCycle™
LSR10040B525 |
25 x 5 cm | 1 Pk. | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
01-182-085
|
SiliCycle™
LSR10040B5253 |
25 x 5 cm | 3 Pk. | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
01-182-086
|
SiliCycle™
LSR10040B5255 |
25 x 5 cm | 5 Pk. | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
01-182-084
|
SiliCycle™
LSR10040B52510 |
25 x 5 cm | 10 Pk. | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Suitable for very dirty purification
- Easier to handle
- Suitable for organic molecules (MW <1000 Dalton)
- E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
- Testing has shown a good correlation between the performance of lab, pilot, and commercial size cartridges.
- Lab cartridges can be operated with a peristaltic pump or a small pressure vessel.
Specifications
Purification Cartridge | |
60 to 200 μm | |
60 Å | |
75 g |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title