Learn More
SiliCycle™ E-PAK™ Cartridges, Pilot Scale, SiliaCarb™ C-HA Activated Carbon
E-PAK™ Cartridges, Pilot Scale, SiliaCarb™ C-HA Activated Carbon is a high-activity wood based powdered activated carbon conveniently packaged in a cartridge.
Supplier:  SiliCycle™ PSCHA1612
 View more versions of this product
Catalog No. 01-182-132
Add to cart
Description
Description
  • Testing with loose media can be done with samples as small as a few milliliters and are normally done in advance of cartridge testing to identify the formula with the highest capacity to remove contaminants with the least loss of product.
  • Testing has shown good correlation between the performance of lab, pilot and commercial size cartridges.
Specifications
Specifications
Pilot Scale E-PAK Cartridge
Black
1 Pk.
Highly Active Carbon
Removal of precious metal catalysts, pigments and reaction contaminants
12 x 16.5 cm
6 to 8
850 g