Learn More
SiliCycle™ E-PAK™ Cartridges, Pilot Scale, SiliaMetS™ Diamine
E-PAK™ SiliaMetS™ Diamine is effective for scavenging acids, acid chlorides, anhydrides, aldehydes, isocyanates, and chloroformates as well as Pb, Ni, and Cd conveniently packaged in a cartridge.
Supplier:  SiliCycle™ PSR49030B1625
 View more versions of this product
Catalog No. 01-182-141
Add to cart
Description
Description
Metals removed:
  • Best scavenger: Cr, Pd, Pt, W, and Zn
  • Good scavenger: Cd, Co, Cu, Fe, Hg, Ni, Pb, Ru, Sc, Se, and V
Additional Information:
  • To facilitate the evaluations of small samples, we can provide loose media and lab scale cartridges on a custom basis.
  • Testing with loose media can be done with samples as small as a few milliliters and is normally done in advance of cartridge testing to identify the formula with the highest capacity to remove contaminants with the least loss of product.
Specifications
Specifications
Pilot Scale E-PAK Cartridge
White to yellow
1 Pk.
40 to 63 μm
1.7 kg
Lower residual metal concentration in Cr, Pd, Pt, W, Zn
25 x 16.5 cm
2 to 9
Diamine