Learn More
SiliCycle™ E-PAK™ Cartridges, Pilot Scale, SiliaMetS™ DMT
E-PAK™ Cartridges, Pilot Scale, SiliaMetS™ DMT (2,4,6-trimercaptotriazine) is a silica-bound scavenger for a variety of metals in particular ruthenium catalysts and hindered Pd complexes conveniently packaged in a cartridge.
Supplier:  SiliCycle™ PSR79030B1612
 View more versions of this product
Catalog No. 01-182-144
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Metals removed:
  • Best scavenger: Ir, Ni, Os, Pd, Pt, Rh, and Ru
  • Good scavenger: Cd, Co, Cu, Fe, Sc, and Zn
Additional Information:
  • To facilitate the evaluations of small samples, we can provide loose media and lab scale cartridges on a custom basis.
  • Testing with loose media can be done with samples as small as a few milliliters and are normally done in advance of cartridge testing to identify the formula with the highest capacity to remove contaminants with the least loss of product.
  • Testing has shown good correlation between the performance of lab, pilot and commercial size cartridges.
Specifications
Specifications
Pilot Scale E-PAK Cartridge
White to yellow
1 Pk.
40 to 63 μm
850 g
Lower residual metal concentration in Ir, Ni, Os, Pd, Pt, Rh and Ru
12 x 16.5 cm
2 to 9
DMT