Learn More
SiliCycle™ E-PAK™ Cartridges, Pilot Scale, SiliaMetS™ Thiol
E-PAK™ Cartridges, Pilot Scale, SiliaMetS™ Thiol (Si-Thiol) is our most versatile and robust metal scavenger for a variety of metals under a wide range of conditions conveniently packaged in a cartridge.
Supplier:  SiliCycle™ PSR51030B1625
 View more versions of this product
Catalog No. 01-182-143
Add to Cart
Description
Description
Metals removed:
  • Best scavenger: Ag, Hg, Os, Pd, and Ru
  • Good scavenger: Cu, Ir, Pb, Rh, Se, and Sn
Additional Information:
  • To facilitate the evaluations of small samples, we can provide loose media and lab scale cartridges on a custom basis.
  • Testing with loose media can be done with samples as small as a few milliliters and is normally done in advance of cartridge testing to identify the formula with the highest capacity to remove contaminants with the least loss of product.
  • Testing has shown a good correlation between the performance of lab, pilot, and commercial size cartridges.
Specifications
Specifications
Pilot Scale E-PAK Cartridge
White to yellow
1 Pk.
40 to 63 μm
1.7 kg
Lower residual metal concentration in Ag, Hg, Os, Pd, Ru
25 x 16.5 cm
2 to 9
Thiol