Learn More
SiliCycle™ E-PAK™ Cartridges, SiliaMetS™ Diamine
E-PAK™ Cartridges, SiliaMetS™ Diamine (Si-DIA) is effective for scavenging acids as well as Pb, Ni, and Cd, conveniently packaged in a cartridge. E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
Supplier:  SiliCycle™ LSR49030B5103
 View more versions of this product
Catalog No. 01-182-092
Add to cart
Description
Description
Metals removed:
  • Best scavenger: Cr, Pd, Pt, W, and Zn
  • Good scavenger: Cd, Co, Cu, Fe, Hg, Ni, Pb, Ru, Sc, Se, and V
Additional Information:
  • To facilitate the evaluations of small samples, we can provide loose media and lab scale cartridges on a custom basis.
  • Testing with loose media can be done with samples as small as a few milliliters and is normally done in advance of cartridge testing to identify the formula with the highest capacity to remove contaminants with the least loss of product.
  • E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
  • Testing has shown a good correlation between the performance of lab, pilot, and commercial size cartridges.
  • Lab cartridges can be operated with a peristaltic pump or a small pressure vessel.
Specifications
Specifications
Lab Scale E-PAK Cartridge
White to yellow
3 Pk.
40 to 63 μm
75 g
Lower residual metal concentration in Cr, Pd, Pt, W, Zn
10 x 5 cm
2 to 9
Diamine