Learn More
SiliCycle™ E-PAK™ Cartridges, SiliaMetS™ DMT
E-PAK™ Cartridges, SiliaMetS™ DMT (2,4,6-trimercaptotriazine) is a silica-bound scavenger for a variety of metals, in particular, ruthenium catalysts and hindered Pd complexes conveniently packaged in a cartridge. E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
Supplier:  SiliCycle™ LSR79030B5110
 View more versions of this product
Catalog No. 01-182-121
Add to cart
Description
Description
Metals removed:
  • Best scavenger: Ir, Ni, Os, Pd, Pt, Rh, and Ru
  • Good scavenger: Cd, Co, Cu, Fe, Sc, and Zn
Additional Information:
  • To facilitate the evaluations of small samples, we can provide loose media and lab scale cartridges on a custom basis.
  • Testing with loose media can be done with samples as small as a few milliliters and are normally done in advance of cartridge testing to identify the formula with the highest capacity to remove contaminants with the least loss of product.
  • E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
  • Testing has shown good correlation between the performance of lab, pilot and commercial size cartridges.
  • Lab cartridges can be operated with pump or small pressure vessel.
Specifications
Specifications
Lab Scale E-PAK Cartridge
White to yellow
10 Pk.
40 to 63 μm
8 g
Lower residual metal concentration in Ir, Ni, Os, Pd, Pt, Rh and Ru
1 x 5 cm
2 to 9
DMT