Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ E-Selectin/CD62E Antibody Pair [HRP]

Rabbit Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2796335PLATES

 View more versions of this product

Catalog No. NB010306


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rat E-Selectin/CD62E.
Specifications

Specifications

HRP
Antibody Pairs
18.8 pg/mL
6401
RUO
E-Selectin/CD62E
Immunology, Lipid and Metabolism
Rat
SELE
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1200 pg/mL is recommended.
Sandwich ELISA
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rat E-Selectin/CD62E.
Sandwich ELISA
  • Detection Antibody: 0.5 mg/mL of rabbit anti-E-Selectin/CD62E monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Capture Antibody: 0.4 mg/mL of rabbit anti-Rat E-Selectin/CD62E monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 33 ng of recombinant Rat E-Selectin/CD62E.
  • Storage is content dependent.
    Rabbit
    5 Plates
    18.8-1200 pg/mL
    CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E
    Conjugated
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit