Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Selectin/CD62E Rabbit anti-Human, Alexa Fluor 594, Clone: 103, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289431AF594

Catalog No. NB066834


Add to cart

Description

Description

E-Selectin/CD62E Monoclonal antibody specifically detects E-Selectin/CD62E in Human samples. It is validated for Flow Cytometry, ELISA
Specifications

Specifications

E-Selectin/CD62E
103
Flow Cytometry, ELISA
Purified
Rabbit
IgG
0.1 mL
Immunology, Lipid and Metabolism
Primary
6401
ELISA, Flow Cytometry
Alexa Fluor 594
50mM Sodium Borate
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E
Recombinant Human E-Selectin/CD62E protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit