Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E-Selectin/CD62E Rabbit anti-Human, Clone: 208, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | E-Selectin/CD62E |
---|---|
Clone | 208 |
Dilution | Immunocytochemistry/Immunofluorescence 1:20-1:100 |
Applications | Immunofluorescence |
Classification | Monoclonal |
Description
E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
E-Selectin/CD62E | |
Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E | |
Recombinant Human E-Selectin/CD62E Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
208 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Immunology, Lipid and Metabolism | |
0.2 um filtered solution in PBS | |
6401 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title