Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Selectin/CD62E Rabbit anti-Mouse, DyLight 650, Clone: 15, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290702C

Catalog No. NB040733


Add to cart

Description

Description

E-Selectin/CD62E Monoclonal antibody specifically detects E-Selectin/CD62E in Mouse samples. It is validated for Western Blot
Specifications

Specifications

E-Selectin/CD62E
15
Western Blot
Purified
Rabbit
IgG
0.1 mL
Immunology, Lipid and Metabolism
Primary
6401
Western Blot
DyLight 650
50mM Sodium Borate
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E
Recombinant Mouse E-Selectin/CD62E Protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit