Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E-Selectin/CD62E Rabbit, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
$551.00
Specifications
Antigen | E-Selectin/CD62E |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5-4 ug/ml, Simple Western 1:50, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 10-20 ug/ml, Immunohistochemistry-Paraffin 10-20 ug/ml |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP145545
|
Novus Biologicals
NBP145545 |
0.1 mg |
Each for $551.00
|
|
NBP145545SS
|
Novus Biologicals
NBP145545SS |
0.025 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
E-Selectin/CD62E Polyclonal specifically detects E-Selectin/CD62E in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
E-Selectin/CD62E | |
Western Blot 0.5-4 ug/ml, Simple Western 1:50, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 10-20 ug/ml, Immunohistochemistry-Paraffin 10-20 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
P16581 | |
6401 | |
Synthetic peptide surrounding amino acid 584 of human E-Selectin. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Immunology, Lipid and Metabolism | |
PBS (pH 7.2), 5 mM EDTA, 0.5% BSA, 30% Glycerol with 0.03% Proclin 300 | |
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E | |
SELE | |
IgG | |
Affinity purified | |
67 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title