Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E-Tag Epitope Tag Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Supplier: Novus Biologicals NB600518B
Description
E-Tag Epitope Tag Polyclonal antibody specifically detects E-Tag Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
E-Tag Epitope Tag | |
Polyclonal | |
PBS | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Purified | |
Epitope Tag |
Western Blot, ELISA, Immunofluorescence | |
Biotin | |
E epitope tag, GAPVPYPDPLEPR epitope tag, GAPVPYPDPLEPR tag | |
Goats were immunized with E-Tag (GAPVPYPDPLEPR) conjugated to KLH. | |
0.1 mL | |
Epitope Tags | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction