Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ E Tag Monoclonal Antibody (11H12B3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA538276
Description
This antibody has been tested in direct-ELISA.
E tag is an epitope tag.
Specifications
E Tag | |
Monoclonal | |
1.27 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Tag | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation, Western Blot | |
11H12B3 | |
Unconjugated | |
E epitope tag; GAPVPYPDPLEPR; GAPVPYPDPLEPR tag | |
GAPVPYPDPLEPRC synthetic peptide conjugate to KLH. | |
100 μL | |
Primary | |
-20°C or -80°C if preferred | |
Liquid | |
Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction