Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ E tag Monoclonal Antibody (B2-E2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA541078
Description
E tag Monoclonal Antibody for Western Blot, ELISA
E tag is an epitope tag.
Specifications
E tag | |
Monoclonal | |
2 mg/mL | |
PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Tag | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
B2-E2 | |
Unconjugated | |
E epitope tag; GAPVPYPDPLEPR; GAPVPYPDPLEPR tag | |
peptide. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction