Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E Tag Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA126544
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA126544 100 μg
1 options

Catalog No. PIPA126544

Supplier: Invitrogen™ PA126544

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-26544 detects E tag.

E tag is an epitope tag.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E Tag
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias E epitope tag; GAPVPYPDPLEPR; GAPVPYPDPLEPR tag
Host Species Rabbit
Immunogen Synthetic peptide: GAPVPYPDPLEPR conjugated to KLH.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.