Learn More
Sklar InstrumentsSupplier Diversity Partner E-Z Lock Instrument Stringer, Sklar, 5 in. x 6 in., Stainless Steel.EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 101826
E-Z Lock Instrument Stringer, Sklar, 5 in. x 6 in., Stainless Steel. Sklar's E-Z Locking Instrument Stringers are used to bundle instruments for sterilization. It is designed to keep instruments in the open position in order to access all debris remaining after a procedure. The rectangular bar has one hinged end that opens to accommodate ringed instruments.
Catalog No. 50-143-7139