Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ EDDP perchlorate Solution, 1.0 mg/mL in Methanol (as pyrolinium), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco E0221ML
Description
A certified Snap-N-Spike™ solution of the major urinary metabolite of methadone. Methadone, sold under various trade names such as Symoron, Amidone or Methadose™, is a synthetic opioid used medically as an analgesic and as a maintenance anti-addictive for use in patients with opioid dependency. Suitable uses for this reference standard include as starting material in preparation of calibrators and controls for LC/MS or GC/MS methods in methadone testing for applications in clinical toxicology or forensic analysis.Specifications
67-56-1, 31161-17-8 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
Methadone metabolite | |
Ampule | |
9.7°C (49.5°F) - closed cup |
C20H24ClNO4 | |
1.0 mg/mL in methanol (as pyrolinium) | |
377.86 | |
1 mL |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?
Provide Content Correction