Learn More
Thermo Scientific™ E1-ClipTip™ Bluetooth™ Electronic Adjustable Tip Spacing Multichannel Equalizer Pipettes
Perform sample transfers between virtually any tube, rack, microplate or horizontal gel box quickly and efficiently with adjustable tip spacing.
Manufacturer:  Thermo Scientific™ 4672040
 View more versions of this product
Catalog No. 14-387--973
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Perform sample transfers between virtually any tube, rack, microplate or horizontal gel box quickly and efficiently with the Thermo Scientific™ E1-ClipTip™ Equalizer Pipette. Adjustable tip spacing allows you to set the distance between tips by simply sliding the scale to expand or contract to the desired labware format.The NEW My Pipette™ Creator app enables programming of E1-ClipTip from the comfort of your PC.
Specifications
Specifications
±2.0 to 10.0%
ClipTip 384 30
0.01µL
Violet
12
1 to 30µL
4.5 to 9.0
E1-ClipTip Equalizer, Format: 384
≤0.9 to 7.0%
E1-ClipTip
ClipTip 384 and ClipTip 12.5 tips and compatible E1-ClipTip models do not have interlocking technology