Learn More
Thermo Scientific™ E1-ClipTip™ Electronic Single Channel Pipettes
Electronic tip ejection, index finger pipetting action and Thermo Scientitic™ ClipTip™ technology make the Thermo Scientific™ E1-ClipTip™ Electronic Single Channel Pipettes ideal for repetitive pipetting tasks with microcentrifuge or test tubes.
Specifications
Model E1-ClipTip
Number of Channels 1
View More Specs
Products 1
Catalog Number Mfr. No. Number of Channels Volume (Metric) Compatible Tips Price Quantity & Availability  
Catalog Number Mfr. No. Number of Channels Volume (Metric) Compatible Tips Price Quantity & Availability  
14-387--953
View Documents
Thermo Scientific™
4670040
1 15 to 1250 μL ClipTip 1250 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Electronic tip ejection, index finger pipetting action and Thermo Scientitic™ ClipTip™ technology make the Thermo Scientific™ E1-ClipTip™ Electronic Single Channel Pipettes ideal for repetitive pipetting tasks with microcentrifuge or test tubes. Feel the difference.
Innovative ClipTip interlocking tip interface technology
  • Tip is locked and sealed in place until ejected — no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
  • Tips are attached and ejected with a light touch — eliminates the frustration of banging tips on the pipette
An ideal choice for sample transfers
  • Reduce repetition and save time between labware formats with adjustable tip spacing in Equalizer models.
Unified operation for multiple users and environments
  • Personalized user interface and password protection ensure consistent operation among multiple users.
Electronic operation to help you enjoy pipetting again
  • Designed for optimum comfort and ease of use, the electronic tip ejection and index finger pipetting action let your thumb relax during pipetting.
Two operating options ensure optimal function for every application
  • Choose a step based Matrix or Presets with settings for your most common applications
MyPipette™ - One click access for more efficient and unified pipetting
  • Customize your main menu view by organizing icons in your preferred order or creating shortcuts for your favorite programs and pipetting functions. You can also protect important settings from unintentional editing with a password.
Store up to 20 programs
  • Tired of continuous editing? The Programs function allows you to store 20 your most common Matrix or Presets protocols with a specific name.
A pipette for every application
  • Choose from single channels, 8- and 16-channel multichannels, or 6- and 12-channel adjustable tip spacing Equalizer multichannel pipettes within the volume range of 0.5-1250µL
Track your pipette
  • Service and calibration trackers remind you when the pipette needs to be sent to serviced
Liquid specific calibrations
  • Higher quality results with liquid specific calibrations
Li-Ion battery
  • Long lasting performance and fast recharging.
Flexible re-charging
  • Use either charging stand or a plug, you decide
2-year warranty
  • With convenient web registration
Compatible with Thermo Scientific ClipTip pipette tips.
†E1-ClipTip 12.5 and all 384 E1-ClipTip pipettes utilize snap and lip tip fitting technology
Specifications
Specifications
E1-ClipTip
1