Learn More
Thermo Scientific™ E1-ClipTip™ Electronic Single Channel Pipettes
Electronic tip ejection, index finger pipetting action and Thermo Scientitic™ ClipTip™ technology make the Thermo Scientific™ E1-ClipTip™ Electronic Single Channel Pipettes ideal for repetitive pipetting tasks with microcentrifuge or test tubes.
Manufacturer:  Thermo Scientific™ 4670020
 View more versions of this product
Catalog No. 14-387--951
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Electronic tip ejection, index finger pipetting action and Thermo Scientitic™ ClipTip™ technology make the Thermo Scientific™ E1-ClipTip™ Electronic Single Channel Pipettes ideal for repetitive pipetting tasks with microcentrifuge or test tubes. Feel the difference.
Specifications
Specifications
±0.6 to 2.4%
0.1
Yellow
1
ClipTip 200
≤0.2 to 0.8%
E1-ClipTip
2 to 125μL