Thermo Scientific E1-ClipTip™ Electronic Single Channel Pipettes
Electronic tip ejection, index finger pipetting action and ClipTip technology are ideal for repetitive pipetting tasks with microcentrifuge or test tubes.
Manufacturer: Thermo Scientific 4670030
 View more versions of this product
Catalog No. 14-387--952
  Check Availability
Add to Cart
    Specifications
    Model E1-ClipTip™
    Volume (Metric) Range 10 to 300μL
    Precision ≤0.2 to 0.5%
    Number of Channels Single Channel
    Color Code Orange,Orange
    Accuracy ±0.6 to 1.7%
    Compatible Tips ClipTip 300,ClipTip 300
    Increments 0.1
    Description E1-ClipTip Single Channel
    Electronic tip ejection, index finger pipetting action and Thermo Scientitic™ ClipTip™ technology make the Thermo Scientific™ E1-ClipTip™ Electronic Single Channel Pipettes ideal for repetitive pipetting tasks with microcentrifuge or test tubes. Feel the difference.
    Innovative ClipTip interlocking tip interface technology†
    · Tip is locked and sealed in place until ejected — no dropped tips that waste valuable samples
    · Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
    · Tips are attached and ejected with a light touch — eliminates the frustration of banging tips on the pipette
    An ideal choice for sample transfers
    · Reduce repetition and save time between labware formats with adjustable tip spacing in Equalizer models.
    Unified operation for multiple users and environments 
    · Personalized user interface and password protection ensure consistent operation among multiple users.
    Electronic operation to help you enjoy pipetting again
    · Designed for optimum comfort and ease of use, the electronic tip ejection and index finger pipetting action let your thumb relax during pipetting.
    Two operating options ensure optimal function for every application
    • Choose a step based Matrix or Presets with settings for your most common applications 
    MyPipette™ - One click access for more efficient and unified pipetting
    • Customize your main menu view by organizing icons in your preferred order or creating shortcuts for your favorite programs and pipetting functions. You can also protect important settings from unintentional editing with a password.
    Store up to 20 programs 
    • Tired of continuous editing? The Programs function allows you to store 20 your most common Matrix or Presets protocols with a specific name. 
    A pipette for every application
    • Choose from single channels, 8- and 16-channel multichannels, or 6- and 12-channel adjustable tip spacing Equalizer multichannel pipettes within the volume range of 0.5-1250µL
    Track your pipette
    • Service and calibration trackers remind you when the pipette needs to be sent to serviced
    Liquid specific calibrations
    • Higher quality results with liquid specific calibrations 
    Li-Ion battery
    • Long lasting performance and fast recharging.
    Flexible re-charging
    • Use either charging stand or a plug, you decide
    2-year warranty 
    • With convenient web registration
    Compatible with Thermo Scientific ClipTip pipette tips.
    †E1-ClipTip 12.5 and all 384 E1-ClipTip pipettes utilize snap and lip tip fitting technology
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback