Learn More
Thermo Scientific™ E1-ClipTip™ Multichannel Pipettes
With enhanced versatility and customization, the Thermo Scientific™ E1-ClipTip™ Electronic Multichannel Pipette automates daily 96-and 384-well microplate pipetting tasks with the sealed in security of Thermo Scientific™ ClipTip™ technology.
Manufacturer:  Thermo Scientific™ 4671020
 View more versions of this product
Catalog No. 14-387--962
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
With enhanced versatility and customization, the Thermo Scientific™ E1-ClipTip™ Electronic Multichannel Pipette automates daily 96-and 384-well microplate pipetting tasks with the sealed in security of Thermo Scientific™ ClipTip™ technology. Enjoy pipetting again with optimum functionality for every application and user preference. Feel the difference.
Specifications
Specifications
±2.5 to 12.0%
E1-ClipTip 16-channel
≤1.6 to 12.0%
E1-ClipTip
ClipTip 384 and ClipTip 12.5 tips and compatible E1-ClipTip models do not have interlocking technology
ClipTip 384 12.5
0.01µL
Pink
16
0.5 to 12.5µL