Learn More
Thermo Scientific™ E1-ClipTip™ Multichannel Pipettes
With enhanced versatility and customization, the Thermo Scientific™ E1-ClipTip™ Electronic Multichannel Pipette automates daily 96-and 384-well microplate pipetting tasks with the sealed in security of Thermo Scientific™ ClipTip™ technology.
Manufacturer:  Thermo Scientific™ 4671070
 View more versions of this product
Catalog No. 14-387--956
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
With enhanced versatility and customization, the Thermo Scientific™ E1-ClipTip™ Electronic Multichannel Pipette automates daily 96-and 384-well microplate pipetting tasks with the sealed in security of Thermo Scientific™ ClipTip™ technology. Enjoy pipetting again with optimum functionality for every application and user preference. Feel the difference.
Specifications
Specifications
±2.0 to 5.0%
0.1µL
Orange
8
ClipTip 300
≤0.6 to 2.0%
E1-ClipTip
10 to 300µL