Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ E2 (HCV) (AY460204, 383 a.a. - 663 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells

Used for SDS-PAGE

Manufacturer:  Abnova™ P5119

Catalog No. 89-942-667


Add to cart

Specifications

Specifications

AY460204
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
E2 (HCV) Recombinant Protein
≥96% by SDS-PAGE
50 ug
RUO
Mammalian cell (293) expression system
Yes
1 ug/uL
Liquid
32kDa
Mammalian cell (293) expression system
SDS-PAGE Result
Store at -20°C. Stable for 6 month from the date of shipment.
E2
Human
His
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit