Learn More
E2F-1 Mouse anti-Human, Alexa Fluor 405, Clone: 10D1 2B5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB600208AF405
Catalog No. NBX208AF405
Add to cart
Description
Description
E2F-1 Monoclonal antibody specifically detects E2F-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
E2F-1
10D1 2B5
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
E2F1
The region encoding the DP-1 binding site (amino acids 223-332) of E2F-1 expressed in E. coli.
Protein G purified
Apoptosis, Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators
Primary
1869
Human
Western Blot, Immunocytochemistry, Immunofluorescence
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
E2F transcription factor 1, E2F-1, PBR3, PRB-binding protein E2F-1, RBAP1, RBAP-1, RBBP3, RBBP-3, RBP3, Retinoblastoma-associated protein 1, Retinoblastoma-binding protein 3, transcription factor E2F1
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
This is specific for human E2F-1.