Learn More
E2F-1 Mouse anti-Human, Alexa Fluor 488, Clone: 1700, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB600206AF488
Catalog No. NBX206AF488
Add to cart
Description
Description
E2F-1 Monoclonal antibody specifically detects E2F-1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
E2F-1
1700
Western Blot
Purified
E2F1
The transactivation and retinoblastoma protein binding domain (amino acids 332-390) of E2F-1 expressed in E. coli.
Protein G purified
Apoptosis, Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators
Primary
1869
Human
Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
E2F transcription factor 1, E2F-1, PBR3, PRB-binding protein E2F-1, RBAP1, RBAP-1, RBBP3, RBBP-3, RBP3, Retinoblastoma-associated protein 1, Retinoblastoma-binding protein 3, transcription factor E2F1
Mouse
IgG1
100 ul
Store at 4C in the dark.
Monoclonal
This is specific for human E2F-1.