Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ E2F-1 Transcription Factor Ab-6, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and gel supershift experiments.

Specifications

Antigen E2F-1 Transcription Factor Ab-6
Clone KH95
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS879R1
View Documents
Thermo Scientific™
MS879R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and gel supershift experiments with Thermo Scientific™ E2F-1 Transcription Factor Ab-6, Mouse Monoclonal Antibody.

E2F's are DNA-binding proteins that associate with negative regulators, such as the retinoblastoma p107 protein, resulting in an altered rate of gene transcription. E2F-1 also requires DP-1 for efficient DNA-binding and transcription modification. E2F1 is proposed to be involved in several cellular processes that range from tumor suppression, cell cycle progression, and oncogenesis. E2F-1 overexpression can also drive cells into apoptosis. These observations place E2F among a group of apoptosis inducers that includes c-Myc, Adenovirus E1A, and HPV E7 protein.

Host Species: Mouse

Clone: KH95

Isotype: IgG2a/κ

Species Reactivity: Human and Mouse. Does not react with rat. Others not known.

Epitope: aa 342-386

Immunogen: Recombinant human E2F-1 protein

Molecular Weight: ∽60 kDa

Positive Control: HeLa or MAD109 cells, Tonsil or Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

E2F-1 Transcription Factor Ab-6
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Transcription Regulation
Recombinant human E2F-1 protein
KH95
Monoclonal
RUO
Primary
IgG2a, κ
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit