Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F-2 Rabbit anti-Human, Clone: SN201-04, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267723
Description
E2F-2 Monoclonal antibody specifically detects E2F-2 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
E2F-2 | |
SN201-04 | |
Unconjugated | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
E2F transcription factor 2, E2F-2transcription factor E2F2 | |
Rabbit | |
IgG | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human, Mouse, Rat |
Flow Cytometry, Western Blot | |
1 mg/mL | |
Western Blot 1:1000-1:2000, Flow Cytometry 1:50-1:100 | |
Purified | |
E2F2 | |
Recombinant Protein | |
Protein A purified | |
Cell Cycle and Replication, Transcription Factors and Regulators | |
Primary | |
1870 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title