Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E2F-2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP31047825UL

 View more versions of this product

Catalog No. NB125858


Add to cart

Description

Description

E2F-2 Polyclonal antibody specifically detects E2F-2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

E2F-2
Unconjugated
PBS buffer, 2% sucrose
E2F transcription factor 2, E2F-2transcription factor E2F2
The immunogen is a synthetic peptide directed towards the middle region of human E2F-2 (NP_004082). Peptide sequence DQFLSPTLACSSPLISFSPSLDQDDYLWGLEAGEGISDLFDSYDLGDLLI
25 μg
Cell Cycle and Replication, Transcription Factors and Regulators
Primary
1870
Western Blot, Immunohistochemistry
Western Blot 1.0 ug/ml, Immunohistochemistry
Purified
Rabbit
Affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit