Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ E2F-4 Transcription Factor Ab-4, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and gel supershift experiments.

Specifications

Antigen E2F-4 Transcription Factor Ab-4
Clone 4E2F04; same as WUF10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1057R1
View Documents
Thermo Scientific™
MS1057R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and gel supershift experiments with Thermo Scientific™ E2F-4 Transcription Factor Ab-4, Mouse Monoclonal Antibody.

E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F-4 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides. Differential phosphorylation contributes to the microheterogeniety of E2F-4 (total aa 411-416, varies due to a trinucleotide repeat).

Host Species: Mouse

Clone: 4E2F04; Same as WUF10

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant human E2F-4 protein

Molecular Weight: ∽56 kDa

Positive Control: Raji cells, Tonsil

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

E2F-4 Transcription Factor Ab-4
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Murine
Transcription Regulation
Recombinant human E2F-4 protein
4E2F04; same as WUF10
Monoclonal
RUO
Primary
IgG1
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit