Learn More
Abnova™ E2F2 293T Cell Transient Overexpression Lysate (Denatured) (T04)
Human overexpression lysate
Manufacturer:  Abnova™ H00001870T04
Catalog No. 89-942-675
Add to cart
Specifications
Specifications
Western Blot
293T
BC053676.1
pCMV-E2F2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
E2F2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 47.5kDa
Transient overexpression cell lysate was tested with anti-E2F2 antibody (Mfr. No. H00001870-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)