Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F3 Monoclonal Antibody (3E2F04 (PG37)), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA511319
Description
MA5-11319 targets E2F-3 Transcription Factor in GS and IF applications and shows reactivity with Human samples. The MA5-11319 immunogen is recombinant human E2F-3 protein.
The protein encoded by this gene is a member of the E2F family of transcription factors. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. This protein and another 2 members, E2F1 and E2F2, have an additional cyclin binding domain. This protein binds specifically to retinoblastoma protein pRB in a cell-cycle dependent manner.
Specifications
E2F3 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
O00716 | |
E2F3 | |
Recombinant human E2F-3 protein | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
ChIP Assay, Gel Shift, Western Blot | |
3E2F04 (PG37) | |
Unconjugated | |
E2F3 | |
DKFZp686C18211; E2F transcription factor 3; E2f3; E2F-3; E2f3a; E2F3b; KIAA0075; MGC104598; RGD1561600; RP1-177P22.2; Transcription factor E2F3 | |
Mouse | |
Protein A | |
RUO | |
1871 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction