Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA595394
Description
Synthetic peptide sequence: 278-320aa, QSCTLDLKLLTEDSENQRLAYVTYQDIRKISGLKDQTVIVVKA. Reconstitution information: Add 0.2 mL of distilled water will yield a concentration of 500 μg/mL.
The protein encoded by this gene is a member of the E2F family of transcription factors. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. This protein and another 2 members, E2F1 and E2F2, have an additional cyclin binding domain. This protein binds specifically to retinoblastoma protein pRB in a cell-cycle dependent manner.Specifications
E2F3 | |
Polyclonal | |
0.2mg sodium phosphate with 0.9MG NaCl, 0.9MG NaCl and 0.05MG sodium azide, 0.05MG sodium azide | |
O00716, O35261 | |
E2F3 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human E2F3, identical to the related mouse sequence. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
E2F3 | |
Transcription factor E2F3; E2F-3; E2F3; KIAA0075 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
13557, 1871, 291105 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title