Learn More
CERILLIANT™ Estrogens CRM in Serum Solution, 0 pg/mL (Sigmatrix Ultra), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco E5003ML
Catalog No. 01-001-325
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Certified Reference Material of estrone and estradiol free serum for clinical research applications by mass spectrometry. Estrogens blank is ready to be used as blank serum matrix for linearity standards or for calibration verification in LC-MS/MS estrogens testing methods. Separate options for individual levels of estrone and estradiol in serum at concentrations ranging from 10 to 1,000 pg/mL for each analyte and 13C- or deuterium labeled internal standards of estrone and estradiol are also available.
Specifications
Specifications
7732-18-5,53-16-7
C18H22O2
3 mL
270.37
Vial
-20°C
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation