Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E6AP/UBE3A Mouse anti-Human, Clone: PCRP-UBE3A-1A2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30717010UG

 View more versions of this product

Catalog No. NB121813


Add to cart

Description

Description

E6AP/UBE3A Monoclonal antibody specifically detects E6AP/UBE3A in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Protein Array
Specifications

Specifications

E6AP/UBE3A
PCRP-UBE3A-1A2
Unconjugated
10 mM PBS with 0.05% BSA
ANCREPVE6AP, AS, CTCL tumor antigen se37-2, E6AP, E6-AP, E6AP ubiquitin-protein ligase, EC 6.3.2.-, FLJ26981, HPVE6A, human papilloma virus E6-associated protein, Human papillomavirus E6-associated protein, Oncogenic protein-associated protein E6-AP, Renal carcinoma antigen NY-REN-54, ubiquitin protein ligase E3A, ubiquitin-protein ligase E3A
Recombinant full-length human E6AP/UBE3A protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Peptide Array
0.2 mg/ml
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Purified
Mouse
IgG1
100 μg
Apoptosis, Transcription Factors and Regulators
Primary
7337
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit