Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EAAT3 Mouse anti-Rat, Clone: 4D6.2, Novus Biologicals™
Mouse Monoclonal Antibody
$177.15 - $666.00
Specifications
Antigen | EAAT3 |
---|---|
Clone | 4D6.2 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml |
Applications | Western Blot, Immunohistochemistry |
Description
EAAT3 Monoclonal specifically detects EAAT3 in Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
EAAT3 | |
1 mg/mL | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
P43005 | |
6505 | |
C-terminus peptide (KDKSDTISFTQTSQF). | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
4D6.2 | |
Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml | |
Monoclonal | |
Mouse | |
Rat | |
EAAC1excitatory amino acid carrier 1, EAAT3excitatory amino acid transporter 3, Excitatory amino-acid carrier 1, Neuronal and epithelial glutamate transporter, Sodium-dependent glutamate/aspartate transporter 3, solute carrier family 1 (neuronal/epithelial high affinity glutamatetransporter, system Xag), member 1, Solute carrier family 1 member 1 | |
SLC1A1 | |
IgG1 | |
Affinity Purified | |
Glutamate Transporter EAAC1. Based on protein sequence the antibody is expected to react with mouse, bovine and rabbit. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title