Learn More
Selleck Chemical LLCSupplier Diversity Partner EAD1 DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S85762MG
EAD1 is a potent autophagy inhibitor with an IC50 of 5.8 μM in the BxPC3 cells. It has antiproliferative activities in lung and pancreatic cancer cells.
Catalog No. 50-194-8288