Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EAF2 Mouse anti-Human, Clone: 14C2-3E11-4G3, MilliporeSigma™

Mouse Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen EAF2
Clone 14C2-3E11-4G3
Concentration 0.5 mg/mL
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABE1872100
View Documents
MilliporeSigma
MABE1872100UL
100 μL
Each for $373.00
Add to cart
 
MABE187225
View Documents
MilliporeSigma
MABE187225UL
25 μL
Each for $145.80
Add to cart
 
Description

Description

EAF2 Monoclonal antibody specifically detects EAF2 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blotting.
Specifications

Specifications

EAF2
0.5 mg/mL
Unconjugated
Mouse
Human
GST-tagged full-length human recombinant ELL-associated factor 2 (EAF2).
Monoclonal
Protein G purified
14C2-3E11-4G3
Immunohistochemistry, Immunoprecipitation, Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1κ
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit