Learn More
Eagle™ Angled-Shelf Cart
Give your staff greater visibility to what's been stowed with the Eagle Angled-Shelf Cart.
Supplier:  Eagle™ M1836C4
 View more versions of this product
Catalog No. 19-033-820
Add to Cart
Description
Description
Open-wire, angled shelves combine improved ergonomics with increased visibility and flexibility
Four durable chrome shelves may be adjusted, and their angle varied, in 1 in. (2.5cm) increments, to accommodate unusual configurations. Two of the four swivel casters have brakes, and each has a weight capacity of 200 lb. (91kg), ensuring reliable mobility even with a heavy load. Measures 62 7/8 in. high (159cm) and 18 in. (46cm) long. Available with shelf widths of 36 or 48 in. (91 or 122cm).
Specifications
Specifications
Visual Merchandizing Cart
45.72 cm
72 in.
Four open wire angled-shelves and four 1 in. dia. 16G Steel posts, two 5 in. dia. die-casting swivel casters and two resilient wheel with brake (200 lb. weight capacity) , three-sided truss reinforcement braces, 3.5 in. dia. donut bumper
27.2 kg
36 in.
18 in.
Ideal for displaying point of sale or special sale items
182.88 cm
Chrome
60 lb.
91.44 cm
Recommendations
Recommendations