Eagle™ Drum-Containment Pallets and Platforms

Grating removes easily for cleaning

Manufacturer: EAGLE MANUFACTURING 1645

 View more versions of this product

Catalog No. 18-999-4470

  Check Availability
Add to Cart

    Specifications

    Capacity (English) 66 gal.
    Capacity 66 gal., 250L
    Certifications/Compliance EPA, 40 CFR 264.175
    Color Yellow
    Depth (English) 51.5 in.
    Depth (Metric) 131cm
    Height (Metric) 20.32cm
    Load Bearing Capacity (Metric) 3629kg
    Weight (English) 67 lb.
    Width (English) 51.5 in.
    Width (Metric) 131cm
    Material HDPE
    Capacity (Metric) 250L
    For Use With (Application) Contain spills and overflows of hazardous materials, keeping floors contamination
    Height (English) 8 in.
    Load Bearing Capacity (English) 8000 lb.
    Shape Square
    Weight (Metric) 30kg
    Product Type Drum Pallet

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit