Eagle™ Drum-Containment Pallets and Platforms

Grating removes easily for cleaning

Manufacturer: EAGLE MANUFACTURING 1620

 View all options for this product

Catalog No. 18-002

  Check Availability
Add to Cart


    Capacity (English) 66 gal.
    Capacity (Metric) 250L
    Certifications/Compliance EPA, 40 CFR 264.175
    Color Yellow
    Depth (English) 26.25 in.
    Material HDPE
    Depth (Metric) 67cm
    Product Type Drum Pallet
    For Use With (Application) Contain spills and overflows of hazardous materials, keeping floors contamination
    Height (English) 13.75 in.
    Height (Metric) 35cm
    Load Bearing Capacity (English) 4000 lb.
    Load Bearing Capacity (Metric) 1814kg
    Shape Square
    Weight (English) 52 lb.
    Weight (Metric) 24kg
    Width (English) 51 in.
    Width (Metric) 130cm
    Capacity 66 gal., 250L

    Pallets and platforms contain spills and overflows of hazardous materials, keeping floors contamination-free.

    • Solid polyethylene construction provides excellent chemical resistance
    • Consist of translucent tub in “safety yellow,” and black grating with 1.75 in. (4.4cm) square openings
    • Pallet units only are forkliftable
    • Poly ramp can be used with 4-drum inline platform, and with 6-drum and 8-drum low-profile platforms

  • Description & Specifications
  • Accessories

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit