Eagle™ Drum Storage Cabinet

The Eagle Drum Storage Cabinet is designed to let you store 55-gallon drums containing hazardous waste or flammables.

Manufacturer: Eagle™ HAZ1926

 View more versions of this product

Catalog No. 18-887A

  Check Availability
Add to Cart

    Specifications

    No. of Doors 2
    No. of Shelves 1
    Height (English) Interior 62 in.
    Height (Metric) Interior 157.4cm
    Width (English) Interior 28.25 in.
    Width (Metric) Interior 71.75cm
    Height (English) 65 in.
    Height (Metric) 165.1cm
    Width (English) 31.25 in.
    Width (Metric) 79.3cm
    Weight (English) 350 lb.
    Weight (Metric) 158.7kg
    Depth (English) 31.25 in.
    Depth (English) Interior 28.25 in.
    Type Vertical drum storage cabinet
    Depth (Metric) 79.37cm
    Depth (Metric) Interior 71.75cm
    Door Style Manual Close
    For Use With (Application) Flammables
    Includes Drum supports and built-in grounding connectors, drum rollers, one shelf, hazardous waste labels
    Capacity (English) 55 gal.
    Material Steel
    Color Yellow
    Certifications/Compliance FM, NFPA, OSHA
    Dimensions (D x W x H) Interior 28.25 x 28.25 x 62 in. (71.75 x 71.75 x 157.4cm)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit