Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eagle™ Flammables Stackable Cabinet
Eagle Flammables Stackable Cabinet can be placed atop floor model cabinets for additional storage of flammables.
Supplier: Eagle™ ADD14X
Description
- Constructed to the same high-quality specifications as standard-size cabinets
- Ideal for storage of smaller containers of flammables on counter tops, workbenches or on the floor
- Available with two doors, in both manual and self-closing styles
- Fusible link on self-closing doors
Compliance
Meet NFPA Code 30 and OSHA requirements.
Specifications
3 | |
15 gal. | |
48.8 cm | |
46 cm | |
109 cm | |
68.9 kg | |
15 x 40 x 19.25 in. (38 x 101.6 x 48.8 cm) | |
22.25 in. | |
152 lb. | |
OSHA, NFPA | |
19.25 in. | |
15 in. | |
Manual-close |
2 | |
Storage Cabinet | |
57 cm | |
101.6 cm | |
38 cm | |
18 x 43 x 22.25 in. (46 x 109 x 57 cm) | |
Steel | |
43 in. | |
Three adjustable shelve of 14.75 in. D, grounding attachments, three point key-lock on doors, 5.1cm raised, leakproof door sills and four leveling feet, fusible link | |
40 in. | |
18 in. | |
Flammables | |
Yellow |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction